CAS RN 4080-98-2
IUPAC Name:
dodecanedihydrazide
Synonyme:
dodecanedihydrazide
Molekulargewicht (g/mol):
258.37
Summenformel:
C12H26N4O2
InChi Key:
GRGBENNNGZARRZ-UHFFFAOYSA-N
SMILES:
NNC(=O)CCCCCCCCCCC(=O)NN