CAS RN 449808-64-4
CAS RN 449808-64-4
IUPAC Name:
—
Synonyme:
—
Molekulargewicht (g/mol):
—
Summenformel:
—
InChi Key:
—
SMILES:
—
1
–
1
von
1
Ergebnisse
1
R1487 Hydrochloride, MedChemExpress
MedChemExpress R1487 Hydrochloride is a highly potent and selective p38α inhibitor, with Kd values of 0.2 nM and 29 nM for p38α and p38β, respectively.