CAS RN 4506-66-5
CAS RN 4506-66-5
IUPAC Name:
tetrahydrogen benzene-1,2,4,5-tetramine tetrachloride
Synonyme:
tetrahydrogen benzene-1,2,4,5-tetramine tetrachloride
Molekulargewicht (g/mol):
284.01
Summenformel:
C6H14Cl4N4
InChi Key:
BZDGCIJWPWHAOF-UHFFFAOYSA-N
SMILES:
[H+].[H+].[H+].[H+].[Cl-].[Cl-].[Cl-].[Cl-].NC1=CC(N)=C(N)C=C1N
1
–
3
von
3
Ergebnisse
1
Tocris Bioscience™ FAK Inhibitor 14
CAS: 4506-66-5 Summenformel: C6H14Cl4N4
2
FAK Inhibitor 14, TRC
CAS: 4506-66-5 Summenformel: C6H10N4 . 4(HCl) Molekulargewicht (g/mol): 284 Synonym: 1,2,4,5-Benzenetetramine Tetrahydrochloride,1,2,4,5-Benzenetetramine, tetrahydrochloride (6CI,7CI,8CI,9CI),NSC 667249,Y 15 SMILES: Cl.Cl.Cl.Cl.Nc1cc(N)c(N)cc1N
3
Y15, MedChemExpress
MedChemExpress Y15 is a potent and specific inhibitor of focal adhesion kinase (FAK) that inhibits its autophosphorylation activity, decreases the viability of cancer cells, and blocks tumor growth.