CAS RN 459168-41-3
CAS RN 459168-41-3
IUPAC Name:
5-chloro-2-(4-methylpiperazine-1-carbonyl)-1H-indole
Synonyme:
5-chloro-2-(4-methylpiperazine-1-carbonyl)-1H-indole
Molekulargewicht (g/mol):
277.75
Summenformel:
C14H16ClN3O
InChi Key:
HUQJRYMLJBBEDO-UHFFFAOYSA-N
SMILES:
CN1CCN(CC1)C(=O)C1=CC2=CC(Cl)=CC=C2N1
1
–
2
von
2
Ergebnisse
1
Tocris Bioscience™ JNJ 7777120
Selective H4 antagonist
2
JNJ-7777120, MedChemExpress
MedChemExpress JNJ-7777120 is a selective H4R antagonist with Ki of 4 ±1 nM, exhibits >1000-fold selectivity over the other histamin receptors.