CAS RN 497-19-8
Natriumcarbonat, wasserfrei, hochrein, SLR, Fisher Chemical™
CAS: 497-19-8 Summenformel: CNa2O3 Molekulargewicht (g/mol): 105.99 MDL-Nummer: MFCD00003494 InChI-Schlüssel: CDBYLPFSWZWCQE-UHFFFAOYSA-L Synonym: sodium carbonate,disodium carbonate,soda ash,carbonic acid disodium salt,calcined soda,sodium carbonate, anhydrous,carbonic acid, disodium salt,washing soda,sodiumcarbonate,solvay soda PubChem CID: 10340 ChEBI: CHEBI:29377 SMILES: [Na+].[Na+].[O-]C([O-])=O
Natriumcarbonat, 99.8 %, zur Analyse, wasserfrei, Pulver, Thermo Scientific Chemicals
CAS: 497-19-8 Summenformel: CNa2O3 Molekulargewicht (g/mol): 105.99 MDL-Nummer: MFCD00003494 InChI-Schlüssel: CDBYLPFSWZWCQE-UHFFFAOYSA-L Synonym: sodium carbonate,disodium carbonate,soda ash,carbonic acid disodium salt,calcined soda,sodium carbonate, anhydrous,carbonic acid, disodium salt,washing soda,sodiumcarbonate,solvay soda PubChem CID: 10340 ChEBI: CHEBI:29377 IUPAC-Name: Dinatriumcarbonat SMILES: [Na+].[Na+].[O-]C([O-])=O
Natriumcarbonat, 99.5%, reinst, wasserfrei, Thermo Scientific Chemicals
CAS: 497-19-8 Summenformel: CNa2O3 Molekulargewicht (g/mol): 105.99 MDL-Nummer: MFCD00003494 InChI-Schlüssel: CDBYLPFSWZWCQE-UHFFFAOYSA-L Synonym: sodium carbonate,disodium carbonate,soda ash,carbonic acid disodium salt,calcined soda,sodium carbonate, anhydrous,carbonic acid, disodium salt,washing soda,sodiumcarbonate,solvay soda PubChem CID: 10340 ChEBI: CHEBI:29377 SMILES: [Na+].[Na+].[O-]C([O-])=O
Natriumcarbonat, ACS-Primärstandard, 99.95 bis 100.05 % (Trockenbasis), Thermo Scientific Chemicals
CAS: 497-19-8 Summenformel: CNa2O3 Molekulargewicht (g/mol): 105.99 MDL-Nummer: MFCD00003494 InChI-Schlüssel: CDBYLPFSWZWCQE-UHFFFAOYSA-L Synonym: sodium carbonate,disodium carbonate,soda ash,carbonic acid disodium salt,calcined soda,sodium carbonate, anhydrous,carbonic acid, disodium salt,washing soda,sodiumcarbonate,solvay soda PubChem CID: 10340 ChEBI: CHEBI:29377 SMILES: [Na+].[Na+].[O-]C([O-])=O
Natriumkarbonat, wasserfrei 99.95 %, Thermo Scientific Chemicals
CAS: 497-19-8 Summenformel: CNa2O3 Molekulargewicht (g/mol): 105.99 MDL-Nummer: MFCD00003494 InChI-Schlüssel: CDBYLPFSWZWCQE-UHFFFAOYSA-L Synonym: sodium carbonate,disodium carbonate,soda ash,carbonic acid disodium salt,calcined soda,sodium carbonate, anhydrous,carbonic acid, disodium salt,washing soda,sodiumcarbonate,solvay soda PubChem CID: 10340 ChEBI: CHEBI:29377 IUPAC-Name: Dinatriumcarbonat SMILES: [Na+].[Na+].[O-]C([O-])=O
Natriumcarbonat, wasserfrei, ACS, 99.5 % min., Thermo Scientific Chemicals
CAS: 497-19-8 Summenformel: CNa2O3 Molekulargewicht (g/mol): 105.99 MDL-Nummer: MFCD00003494 InChI-Schlüssel: CDBYLPFSWZWCQE-UHFFFAOYSA-L Synonym: sodium carbonate,disodium carbonate,soda ash,carbonic acid disodium salt,calcined soda,sodium carbonate, anhydrous,carbonic acid, disodium salt,washing soda,sodiumcarbonate,solvay soda PubChem CID: 10340 ChEBI: CHEBI:29377 SMILES: [Na+].[Na+].[O-]C([O-])=O
Natriumcarbonat, 99.6 %, ACS-Reagenz, wasserfrei, Thermo Scientific Chemicals
CAS: 497-19-8 Summenformel: CNa2O3 Molekulargewicht (g/mol): 105.99 MDL-Nummer: MFCD00003494 InChI-Schlüssel: CDBYLPFSWZWCQE-UHFFFAOYSA-L Synonym: sodium carbonate,disodium carbonate,soda ash,carbonic acid disodium salt,calcined soda,sodium carbonate, anhydrous,carbonic acid, disodium salt,washing soda,sodiumcarbonate,solvay soda PubChem CID: 10340 ChEBI: CHEBI:29377 IUPAC-Name: Dinatriumcarbonat SMILES: [Na+].[Na+].[O-]C([O-])=O
Natriumcarbonat, 98 %, Thermo Scientific Chemicals
CAS: 497-19-8 Summenformel: CNa2O3 Molekulargewicht (g/mol): 105.99 MDL-Nummer: MFCD00003494 InChI-Schlüssel: CDBYLPFSWZWCQE-UHFFFAOYSA-L Synonym: sodium carbonate,disodium carbonate,soda ash,carbonic acid disodium salt,calcined soda,sodium carbonate, anhydrous,carbonic acid, disodium salt,washing soda,sodiumcarbonate,solvay soda PubChem CID: 10340 ChEBI: CHEBI:29377 SMILES: [Na+].[Na+].[O-]C([O-])=O
Natriumcarbonat, Puriss. P.A., ACS Reagenz, Reag. ISO, Reag. Ph. Eur., wasserfrei ≥, 99,8 % (kalkuliert auf die getrocknete Substanz), Solstice
CAS: 497-19-8 Summenformel: CNa2O3 Molekulargewicht (g/mol): 105.99 MDL-Nummer: MFCD00003494 InChI-Schlüssel: CDBYLPFSWZWCQE-UHFFFAOYSA-L Synonym: Natriumcarbonat,Dinatriumkarbonat,Natriumkarbonat,Kohlensäure-Dinatriumsalz,Calcinierter Soda,Natriumcarbonat, wasserfrei,Kohlensäure, Dinatriumsalz,Waschsoda,Solvay-Natron,solvay soda PubChem CID: 10340 ChEBI: CHEBI:29377 SMILES: [Na+].[Na+].[O-]C([O-])=O
Natriumcarbonat, wasserfrei, AR-zertifiziert für die Analyse, Fisher Chemical™
CAS: 497-19-8 Summenformel: CNa2O3 Molekulargewicht (g/mol): 105.99 MDL-Nummer: MFCD00003494 InChI-Schlüssel: CDBYLPFSWZWCQE-UHFFFAOYSA-L Synonym: sodium carbonate,disodium carbonate,soda ash,carbonic acid disodium salt,calcined soda,sodium carbonate, anhydrous,carbonic acid, disodium salt,washing soda,sodiumcarbonate,solvay soda PubChem CID: 10340 ChEBI: CHEBI:29377 SMILES: [Na+].[Na+].[O-]C([O-])=O
Natriumcarbonat, wasserfrei, 99.997 % (Metallbasis), Thermo Scientific Chemicals, Puratronic™
CAS: 497-19-8 Summenformel: CNa2O3 Molekulargewicht (g/mol): 105.99 MDL-Nummer: MFCD00003494 InChI-Schlüssel: CDBYLPFSWZWCQE-UHFFFAOYSA-L Synonym: sodium carbonate,disodium carbonate,soda ash,carbonic acid disodium salt,calcined soda,sodium carbonate, anhydrous,carbonic acid, disodium salt,washing soda,sodiumcarbonate,solvay soda PubChem CID: 10340 ChEBI: CHEBI:29377 SMILES: [Na+].[Na+].[O-]C([O-])=O
Carbonatlösung, 0,5 M Natriumcarbonat, für Ionenchromatographie, H₂O -Matrix, Fisher chemical™
CAS: 497-19-8 Summenformel: CNa2O3 Molekulargewicht (g/mol): 105.99 MDL-Nummer: MFCD00003494 InChI-Schlüssel: CDBYLPFSWZWCQE-UHFFFAOYSA-L SMILES: [Na+].[Na+].[O-]C([O-])=O
Natriumcarbonat, 99+%, zur Analyse, Granulat, Thermo Scientific Chemicals
CAS: 497-19-8 Summenformel: CNa2O3 Molekulargewicht (g/mol): 105.99 MDL-Nummer: MFCD00003494 InChI-Schlüssel: CDBYLPFSWZWCQE-UHFFFAOYSA-L Synonym: sodium carbonate,disodium carbonate,soda ash,carbonic acid disodium salt,calcined soda,sodium carbonate, anhydrous,carbonic acid, disodium salt,washing soda,sodiumcarbonate,solvay soda PubChem CID: 10340 ChEBI: CHEBI:29377 SMILES: [Na+].[Na+].[O-]C([O-])=O
Natriumcarbonat, 99.999 %, (Metallspurenanalyse), Thermo Scientific Chemicals
CAS: 497-19-8 Summenformel: CNa2O3 Molekulargewicht (g/mol): 105.99 MDL-Nummer: MFCD00003494 InChI-Schlüssel: CDBYLPFSWZWCQE-UHFFFAOYSA-L Synonym: sodium carbonate,disodium carbonate,soda ash,carbonic acid disodium salt,calcined soda,sodium carbonate, anhydrous,carbonic acid, disodium salt,washing soda,sodiumcarbonate,solvay soda PubChem CID: 10340 ChEBI: CHEBI:29377 SMILES: [Na+].[Na+].[O-]C([O-])=O