CAS RN 500-62-9
CAS RN 500-62-9
IUPAC Name:
—
Synonyme:
—
Molekulargewicht (g/mol):
—
Summenformel:
—
InChi Key:
—
SMILES:
—
1
–
2
von
2
Ergebnisse
1
Yangonin, TRC
CAS: 500-62-9 Summenformel: C15 H14 O4 Molekulargewicht (g/mol): 258.27 Synonym: 4-Methoxy-6-[(1E)-2-(4-methoxyphenyl)ethenyl]-2H-pyran-2-one,4-Methoxy-6-[β-(p-anisyl)vinyl]-α-pyrone,6-(p-Methoxystyryl)-4-methoxy-α-pyrone; IUPAC-Name: 4-methoxy-6-[(E)-2-(4-methoxyphenyl)ethenyl]pyran-2-one SMILES: COC1=CC(=O)OC(=C1)\C=C\c2ccc(OC)cc2
2
Yangonin, MedChemExpress
MedChemExpress Yangonin exhibits affinity for the human recombinant cannabinoid CB1 receptor with an IC50 and a Ki of 1.79 μM and 0.72 μM, respectively.