CAS RN 500794-88-7
CAS RN 500794-88-7
IUPAC Name:
—
Synonyme:
—
Molekulargewicht (g/mol):
—
Summenformel:
—
InChi Key:
—
SMILES:
—
1
–
1
von
1
Ergebnisse
1
TR antagonist 1, MedChemExpress
MedChemExpress TR antagonist 1 is a high-affinity thyroid hormone receptor (TR) antagonist with IC50s of 36 and 22 nM for TRα and TRβ, respectively.