CAS RN 511295-01-5
IUPAC Name:
(2,3,5,6-tetrafluorophenyl)boronic acid
Synonyme:
2,3,5,6-tetrafluorophenylboronic acid
Molekulargewicht (g/mol):
193.89
Summenformel:
C6H3BF4O2
InChi Key:
SORPNEUIIZJCBF-UHFFFAOYSA-N
SMILES:
OB(O)C1=C(F)C(F)=CC(F)=C1F