CAS RN 512177-83-2
CAS RN 512177-83-2
IUPAC Name:
—
Synonyme:
—
Molekulargewicht (g/mol):
—
Summenformel:
—
InChi Key:
—
SMILES:
—
1
–
1
von
1
Ergebnisse
1
CCR2-RA-[R], MedChemExpress
MedChemExpress CCR2-RA-[R] is an allosteric antagonist of the C-C chemokine receptor type 2 (CCR2) with an IC50 of 103 nM.