CAS RN 56-40-6
Glycin (weiße Kristalle oder kristallines Pulver), Fisher BioReagents™
CAS: 56-40-6 Summenformel: C2H5NO2 Molekulargewicht (g/mol): 75.07 MDL-Nummer: MFCD00008131 InChI-Schlüssel: DHMQDGOQFOQNFH-UHFFFAOYSA-N Synonym: glycine,aminoacetic acid,glycocoll,aminoethanoic acid,glycolixir,glycosthene,aciport,glicoamin,padil,hampshire glycine PubChem CID: 750 ChEBI: CHEBI:15428 IUPAC-Name: 2-Aminoessigsäure SMILES: NCC(O)=O
Thermo Scientific Chemicals Glycin, ≥99 %, zur Analyse
CAS: 56-40-6 Summenformel: C2H5NO2 Molekulargewicht (g/mol): 75.07 MDL-Nummer: MFCD00008131 InChI-Schlüssel: DHMQDGOQFOQNFH-UHFFFAOYSA-N Synonym: glycine,aminoacetic acid,glycocoll,aminoethanoic acid,glycolixir,glycosthene,aciport,glicoamin,padil,hampshire glycine PubChem CID: 750 ChEBI: CHEBI:15428 IUPAC-Name: 2-Aminoessigsäure SMILES: NCC(O)=O
Glycin, Zertifiziert nach AR zur Analyse, Fisher Chemical™
CAS: 56-40-6 Summenformel: C2H5NO2 Molekulargewicht (g/mol): 75.07 MDL-Nummer: MFCD00008131 InChI-Schlüssel: DHMQDGOQFOQNFH-UHFFFAOYSA-N Synonym: glycine,aminoacetic acid,glycocoll,aminoethanoic acid,glycolixir,glycosthene,aciport,glicoamin,padil,hampshire glycine PubChem CID: 750 ChEBI: CHEBI:15428 IUPAC-Name: 2-Aminoessigsäure SMILES: NCC(O)=O
Thermo Scientific Chemicals Glycin, ≥99 %, ACS Reagenz
CAS: 56-40-6 Summenformel: C2H5NO2 Molekulargewicht (g/mol): 75.07 MDL-Nummer: MFCD00008131 InChI-Schlüssel: DHMQDGOQFOQNFH-UHFFFAOYSA-N Synonym: glycine,aminoacetic acid,glycocoll,aminoethanoic acid,glycolixir,glycosthene,aciport,glicoamin,padil,hampshire glycine PubChem CID: 750 ChEBI: CHEBI:15428 IUPAC-Name: 2-Aminoessigsäure SMILES: NCC(O)=O
Glycin, ≥ 99 %, molekularbiologische Qualität, Ultrapure, Thermo Scientific Chemicals
CAS: 56-40-6 Summenformel: C2H5NO2 Molekulargewicht (g/mol): 75.067 MDL-Nummer: MFCD00008131 InChI-Schlüssel: DHMQDGOQFOQNFH-UHFFFAOYSA-N Synonym: glycine,aminoacetic acid,glycocoll,aminoethanoic acid,glycolixir,glycosthene,aciport,glicoamin,padil,hampshire glycine PubChem CID: 750 ChEBI: CHEBI:15428 IUPAC-Name: 2-Aminoessigsäure SMILES: C(C(=O)O)N
Thermo Scientific Chemicals Glycin, 0.2 M-Pufferlösung, pH 2.5
CAS: 56-40-6 Summenformel: C2H5NO2 Molekulargewicht (g/mol): 75.07 MDL-Nummer: MFCD00008131 InChI-Schlüssel: DHMQDGOQFOQNFH-UHFFFAOYSA-N IUPAC-Name: 2-Aminoessigsäure SMILES: NCC(O)=O
Thermo Scientific Chemicals Glycin, 99 %
CAS: 56-40-6 Summenformel: C2H5NO2 Molekulargewicht (g/mol): 75.07 MDL-Nummer: MFCD00008131 InChI-Schlüssel: DHMQDGOQFOQNFH-UHFFFAOYSA-N Synonym: glycine,aminoacetic acid,glycocoll,aminoethanoic acid,glycolixir,glycosthene,aciport,glicoamin,padil,hampshire glycine PubChem CID: 750 ChEBI: CHEBI:15428 IUPAC-Name: 2-Aminoessigsäure SMILES: NCC(O)=O
Thermo Scientific Chemicals Glycin, Elektrophoresequalität, 99.5+ %
CAS: 56-40-6 Summenformel: C2H5NO2 Molekulargewicht (g/mol): 75.07 MDL-Nummer: MFCD00008131 InChI-Schlüssel: DHMQDGOQFOQNFH-UHFFFAOYSA-N Synonym: glycine,aminoacetic acid,glycocoll,aminoethanoic acid,glycolixir,glycosthene,aciport,glicoamin,padil,hampshire glycine PubChem CID: 750 ChEBI: CHEBI:15428 IUPAC-Name: 2-Aminoessigsäure SMILES: NCC(O)=O
Glycin, 98 %, reinst, Thermo Scientific Chemicals
CAS: 56-40-6 Summenformel: C2H5NO2 Molekulargewicht (g/mol): 75.07 MDL-Nummer: MFCD00008131 InChI-Schlüssel: DHMQDGOQFOQNFH-UHFFFAOYSA-N Synonym: glycine,aminoacetic acid,glycocoll,aminoethanoic acid,glycolixir,glycosthene,aciport,glicoamin,padil,hampshire glycine PubChem CID: 750 ChEBI: CHEBI:15428 IUPAC-Name: 2-Aminoessigsäure SMILES: NCC(O)=O
Thermo Scientific Chemicals Glycin, 0.2 M-Pufferlösung, pH 3.0
CAS: 56-40-6 Summenformel: C2H5NO2 Molekulargewicht (g/mol): 75.07 MDL-Nummer: MFCD00008131 InChI-Schlüssel: DHMQDGOQFOQNFH-UHFFFAOYSA-N IUPAC-Name: 2-Aminoessigsäure SMILES: NCC(O)=O
Glycin, kristallin, MP Biomedicals
CAS: 56-40-6 Summenformel: C2H5NO2 Molekulargewicht (g/mol): 75.07 MDL-Nummer: MFCD00008131 InChI-Schlüssel: DHMQDGOQFOQNFH-UHFFFAOYSA-N Synonym: Glyzin,Aminoessigsäure,Glykocoll,Aminoethansäure,Glykosthen,Hampshire Glycin,aciport,glicoamin,padil,hampshire glycine PubChem CID: 750 ChEBI: CHEBI:15428 IUPAC-Name: 2-Aminoessigsäure SMILES: NCC(O)=O
Thermo Scientific Chemicals Glycin, ACS, 98.5+ %
CAS: 56-40-6 Summenformel: C2H5NO2 Molekulargewicht (g/mol): 75.07 MDL-Nummer: MFCD00008131 InChI-Schlüssel: DHMQDGOQFOQNFH-UHFFFAOYSA-N Synonym: glycine,aminoacetic acid,glycocoll,aminoethanoic acid,glycolixir,glycosthene,aciport,glicoamin,padil,hampshire glycine PubChem CID: 750 ChEBI: CHEBI:15428 IUPAC-Name: 2-Aminoessigsäure SMILES: NCC(O)=O
Thermo Scientific Chemicals Glycin, 0.2 M-Pufferlösung, pH 3.5
CAS: 56-40-6 Molekulargewicht (g/mol): 75,067
Glycin, ACS-Gütegrad, ≥ 98.5 %, Thermo Scientific Chemicals
CAS: 56-40-6 Summenformel: C2H5NO2 Molekulargewicht (g/mol): 75.067 MDL-Nummer: MFCD00008131 InChI-Schlüssel: DHMQDGOQFOQNFH-UHFFFAOYSA-N Synonym: glycine,aminoacetic acid,glycocoll,aminoethanoic acid,glycolixir,glycosthene,aciport,glicoamin,padil,hampshire glycine PubChem CID: 750 ChEBI: CHEBI:15428 IUPAC-Name: 2-Aminoessigsäure SMILES: C(C(=O)O)N