CAS RN 57-13-6
Harnstoff, 98+%, Thermo Scientific Chemicals
CAS: 57-13-6 Summenformel: CH4N2O Molekulargewicht (g/mol): 60.056 MDL-Nummer: MFCD00008022 InChI-Schlüssel: XSQUKJJJFZCRTK-UHFFFAOYSA-N Synonym: carbamide,isourea,carbonyldiamide,ureophil,carbonyldiamine,carbamimidic acid,pseudourea,ureaphil,urevert,alphadrate PubChem CID: 1176 ChEBI: CHEBI:48376 IUPAC-Name: Harnstoff SMILES: C(=O)(N)N
Harnstoff, 99.5 %, zur Analyse, Thermo Scientific Chemicals
CAS: 57-13-6 Summenformel: CH4N2O Molekulargewicht (g/mol): 60.06 InChI-Schlüssel: XSQUKJJJFZCRTK-UHFFFAOYSA-N Synonym: carbamide,isourea,carbonyldiamide,ureophil,carbonyldiamine,carbamimidic acid,pseudourea,ureaphil,urevert,alphadrate PubChem CID: 1176 ChEBI: CHEBI:48376 IUPAC-Name: Harnstoff SMILES: C(=O)(N)N
Harnstoff,99%, Thermo Scientific Chemicals
CAS: 57-13-6 MDL-Nummer: MFCD00008022 InChI-Schlüssel: XSQUKJJJFZCRTK-UHFFFAOYSA-N Synonym: carbamide,isourea,carbonyldiamide,ureophil,carbonyldiamine,carbamimidic acid,pseudourea,ureaphil,urevert,alphadrate PubChem CID: 1176 ChEBI: CHEBI:48376 IUPAC-Name: Harnstoff SMILES: C(=O)(N)N
Harnstoff, hochrein, 99%, Thermo Scientific Chemicals
CAS: 57-13-6 Summenformel: CH4N2O Molekulargewicht (g/mol): 60.056 MDL-Nummer: MFCD00008022 InChI-Schlüssel: XSQUKJJJFZCRTK-UHFFFAOYSA-N Synonym: carbamide,isourea,carbonyldiamide,ureophil,carbonyldiamine,carbamimidic acid,pseudourea,ureaphil,urevert,alphadrate PubChem CID: 1176 ChEBI: CHEBI:48376 IUPAC-Name: Harnstoff SMILES: C(=O)(N)N
Harnstoff, ACS, 99.0-100.5 %, Thermo Scientific Chemicals
CAS: 57-13-6 Summenformel: CH4N2O Molekulargewicht (g/mol): 60.056 MDL-Nummer: MFCD00008022 InChI-Schlüssel: XSQUKJJJFZCRTK-UHFFFAOYSA-N Synonym: carbamide,isourea,carbonyldiamide,ureophil,carbonyldiamine,carbamimidic acid,pseudourea,ureaphil,urevert,alphadrate PubChem CID: 1176 ChEBI: CHEBI:48376 IUPAC-Name: Harnstoff SMILES: C(=O)(N)N
Thermo Scientific Chemicals Harnsäure, ≥ 99 %, für die Biochemie
CAS: 57-13-6 Summenformel: CH4N2O Molekulargewicht (g/mol): 60.06 InChI-Schlüssel: XSQUKJJJFZCRTK-UHFFFAOYSA-N Synonym: carbamide,isourea,carbonyldiamide,ureophil,carbonyldiamine,carbamimidic acid,pseudourea,ureaphil,urevert,alphadrate PubChem CID: 1176 ChEBI: CHEBI:48376 IUPAC-Name: Harnstoff SMILES: C(=O)(N)N
Harnstoff, 99.3+%, Thermo Scientific Chemicals
CAS: 57-13-6 Summenformel: CH4N2O Molekulargewicht (g/mol): 60.056 MDL-Nummer: MFCD00008022 InChI-Schlüssel: XSQUKJJJFZCRTK-UHFFFAOYSA-N Synonym: carbamide,isourea,carbonyldiamide,ureophil,carbonyldiamine,carbamimidic acid,pseudourea,ureaphil,urevert,alphadrate PubChem CID: 1176 ChEBI: CHEBI:48376 IUPAC-Name: Harnstoff SMILES: C(=O)(N)N
Harnstoff, 98 %, reinst, Perlen, Thermo Scientific Chemicals
CAS: 57-13-6 Summenformel: CH4N2O Molekulargewicht (g/mol): 60.06 MDL-Nummer: MFCD00008022 InChI-Schlüssel: XSQUKJJJFZCRTK-UHFFFAOYSA-N Synonym: carbamide,isourea,carbonyldiamide,ureophil,carbonyldiamine,carbamimidic acid,pseudourea,ureaphil,urevert,alphadrate PubChem CID: 1176 ChEBI: CHEBI:48376 IUPAC-Name: Harnstoff SMILES: C(=O)(N)N
Harnstoff, ≥99,5 %, molekularbiologischer Gütegrad, Ultra Pure, Thermo Scientific Chemicals
CAS: 57-13-6 Summenformel: CH4N2O Molekulargewicht (g/mol): 60.056 MDL-Nummer: MFCD00008022 InChI-Schlüssel: XSQUKJJJFZCRTK-UHFFFAOYSA-N Synonym: Carbamid,Isoharnstoff,Carbonyldiamid,Ureaphil,Carbonyldiamin,Carbamidinsäure,Pseudoharnstoff,ureaphil,urevert,alphadrate PubChem CID: 1176 ChEBI: CHEBI:48376 IUPAC-Name: Harnstoff SMILES: C(=O)(N)N
Harnstoff, AR-zertifiziert für Analysen, Fisher Chemical™
CAS: 57-13-6 Summenformel: CH4N2O Molekulargewicht (g/mol): 60.056 MDL-Nummer: 8022 InChI-Schlüssel: XSQUKJJJFZCRTK-UHFFFAOYSA-N Synonym: Carbamid,Isoharnstoff,Carbonyldiamid,Ureaphil,Carbonyldiamin,Carbamidinsäure,Pseudoharnstoff,ureaphil,urevert,alphadrate PubChem CID: 1176 ChEBI: CHEBI:48376 IUPAC-Name: Harnstoff SMILES: C(=O)(N)N
Thermo Scientific Chemicals Harnstoff, 99.5 %, für die Molekularbiologie, DNAse, RNase und Protease frei
CAS: 57-13-6 Summenformel: CH4N2O Molekulargewicht (g/mol): 60.06 MDL-Nummer: MFCD00008022 InChI-Schlüssel: XSQUKJJJFZCRTK-UHFFFAOYSA-N Synonym: carbamide,isourea,carbonyldiamide,ureophil,carbonyldiamine,carbamimidic acid,pseudourea,ureaphil,urevert,alphadrate PubChem CID: 1176 ChEBI: CHEBI:48376 IUPAC-Name: Harnstoff SMILES: C(=O)(N)N
Harnstoff (Farblose bis weiße Kristalle oder kristallines Pulver/Mol. Biol.), Fisher BioReagents™
CAS: 57-13-6 Summenformel: CH4N2O Molekulargewicht (g/mol): 60.056 InChI-Schlüssel: XSQUKJJJFZCRTK-UHFFFAOYSA-N Synonym: carbamide,isourea,carbonyldiamide,ureophil,carbonyldiamine,carbamimidic acid,pseudourea,ureaphil,urevert,alphadrate PubChem CID: 1176 ChEBI: CHEBI:48376 IUPAC-Name: Harnstoff SMILES: C(=O)(N)N
Harnstoff, ultrarein, 5 %, MP Biomedicals
Harnstoff, ≥99 %, (kristallin/ACS-Reagenzienqualität), MP Biomedicals
CAS: 57-13-6 Summenformel: CH4N2O Molekulargewicht (g/mol): 60.056 InChI-Schlüssel: XSQUKJJJFZCRTK-UHFFFAOYSA-N Synonym: Carbamid,Isoharnstoff,Carbonyldiamid,Ureaphil,Carbonyldiamin,Carbamidinsäure,Pseudoharnstoff,ureaphil,urevert,alphadrate PubChem CID: 1176 ChEBI: CHEBI:48376 IUPAC-Name: Harnstoff SMILES: C(=O)(N)N