CAS RN 58020-43-2
CAS RN 58020-43-2
IUPAC Name:
—
Synonyme:
—
Molekulargewicht (g/mol):
—
Summenformel:
—
InChi Key:
—
SMILES:
—
1
–
1
von
1
Ergebnisse
1
HOKU-81, MedChemExpress
MedChemExpress HOKU-81 (4-Hydroxytulobuterol) is one of the metabolites of Tulobuterol (HY-B1810). HOKU-81 is a potent and selective β2-adrenoceptor stimulant. HOKU-81 has bronchodilating effect.