CAS RN 600-18-0
IUPAC Name:
2-oxobutanoic acid
Synonyme:
2-oxobutanoic acid
Molekulargewicht (g/mol):
102.09
Summenformel:
C4H6O3
InChi Key:
TYEYBOSBBBHJIV-UHFFFAOYSA-N
SMILES:
CCC(=O)C(O)=O
1
–
2
von
2
Ergebnisse
1
Alpha-Ketobuttersäure, TRC
CAS: 600-18-0 Summenformel: C4 H6 O3 Molekulargewicht (g/mol): 102.09 Synonym: 2-Oxobutanoic Acid,2-Oxobutyric Acid,2-Ketobutanoic Acid,2-Ketobutyric Acid,2-Oxo-n-butyric Acid,2-Oxobutanoic Acid,2-Oxobutyric Acid,3-Methylpyruvic Acid,NSC 60533,Propionylformic Acid,α-Ketobutyric Acid,α-Oxo-n-butyric Acid,α-Oxobutanoic Acid,α-Oxobutyric Acid,α-keto-n-Butyric Acid IUPAC-Name: 2-Oxobutansäure SMILES: CCC(=O)C(=O)O
2
2-Oxobutanoic acid, MedChemExpress
MedChemExpress 2-Oxobutanoic acid is a product in the enzymatic cleavage of cystathionine.