CAS RN 631-01-6
CAS RN 631-01-6
IUPAC Name:
—
Synonyme:
—
Molekulargewicht (g/mol):
—
Summenformel:
—
InChi Key:
—
SMILES:
—
1
–
2
von
2
Ergebnisse
1
Quillaic Acid, TRC
CAS: 631-01-6 Summenformel: C30H46O5 Molekulargewicht (g/mol): 486.68 Synonym: 3β,16α-Dihydroxy-23-oxo-olean-12-en-28-oic Acid,Quillaja Sapogenin IUPAC-Name: (4aR,5R,6aR,6aS,6bR,8aR,9S,10S,12aR,14bS)-9-formyl-5,10-dihydroxy-2,2,6a,6b,9,12a-hexamethyl-1,3,4,5,6,6a,7,8,8a,10,11,12,13,14b-tetradecahydropicene-4a-carboxylic acid SMILES: [Na+].[Na+].Nc1cc(S(=O)(=O)[O-])c(cc1)CCc2c(cc(N)cc2)S(=O)(=O)[O-]
2
Quillaic acid, MedChemExpress
MedChemExpress Quillaic acid(Quillaja sapogenin) is the major aglycone of the widely studied saponins of the Chilean indigenous tree Quillaja saponaria Mol; can elicit dose-dependent antinociceptive effects in two murine thermal models.