CAS RN 646-01-5
IUPAC Name:
3-(methylsulfanyl)propanoic acid
Synonyme:
3-methylthiopropionic acid 3-methyl-thiopropionic acid
Molekulargewicht (g/mol):
120.17
Summenformel:
C4H8O2S
InChi Key:
CAOMCZAIALVUPA-UHFFFAOYSA-N
SMILES:
CSCCC(O)=O
1
–
2
von
2
Ergebnisse
1
3-(Methylthio)propionsäure, 98 %, Thermo Scientific Chemicals
CAS: 646-01-5 Summenformel: C4H8O2S Molekulargewicht (g/mol): 120.166 MDL-Nummer: MFCD00059635 InChI-Schlüssel: CAOMCZAIALVUPA-UHFFFAOYSA-N Synonym: 3-methylthio propionic acid,3-methylthiopropionate,3-methylthiopropionic acid,4-thiapentanoic acid,3-methylsulfanyl propanoic acid,propanoic acid, 3-methylthio,3-methylmercaptopropionic acid,unii-f523ali47d,3-methylthio propanoic acid,propionic acid, 3-methylthio PubChem CID: 563 ChEBI: CHEBI:1438 IUPAC-Name: 3-Methylsulfanylpropansäure SMILES: CSCCC(=O)O
2
3-(Methylthio)propionic acid, MedChemExpress
MedChemExpress 3-(Methylthio)propionic acid is an intermediate in the methionine metabolism.