CAS RN 694433-59-5
CAS RN 694433-59-5
IUPAC Name:
2-[4-(2H-1,3-benzodioxol-5-yl)-2-tert-butyl-1H-imidazol-5-yl]-6-methylpyridine
Synonyme:
2-[5-(2H-1,3-benzodioxol-5-yl)-2-tert-butyl-3H-imidazol-4-yl]-6-methylpyridine
Molekulargewicht (g/mol):
335.41
Summenformel:
C20H21N3O2
InChi Key:
WGZOTBUYUFBEPZ-UHFFFAOYSA-N
SMILES:
CC1=CC=CC(=N1)C1=C(N=C(N1)C(C)(C)C)C1=CC2=C(OCO2)C=C1
1
–
2
von
2
Ergebnisse
1
Tocris Bioscience™ SB 505124
CAS: 694433-59-5 Summenformel: C20H21N3O2
2
SB-505124, MedChemExpress
MedChemExpress SB-505124 is a selective inhibitor of TGF-β Receptor type I receptors (ALK4, ALK5, ALK7), with IC50s of 129 nM and 47 nM for ALK4, ALK5, respectively, but it does not inhibit ALK1, 2, 3, or 6.