CAS RN 71195-85-2
IUPAC Name:
2,3,4,5,6-pentafluorophenyl prop-2-enoate
Synonyme:
2,3,4,5,6-pentafluorophenyl prop-2-enoate
Molekulargewicht (g/mol):
238.11
Summenformel:
C9H3F5O2
InChi Key:
RFOWDPMCXHVGET-UHFFFAOYSA-N
SMILES:
FC1=C(F)C(F)=C(OC(=O)C=C)C(F)=C1F