CAS RN 736048-65-0
CAS RN 736048-65-0
IUPAC Name:
—
Synonyme:
—
Molekulargewicht (g/mol):
—
Summenformel:
—
InChi Key:
—
SMILES:
—
1
–
1
von
1
Ergebnisse
1
PKC-theta inhibitor, MedChemExpress
MedChemExpress PKC-theta inhibitor is a selective PKC-θinhibitor, with an IC50 of 12 nM.