CAS RN 7440-67-7
Zirkoniumschwamm, 99.5 %, Zr und Hf, Thermo Scientific Chemicals
CAS: 7440-67-7 Summenformel: Zr Molekulargewicht (g/mol): 91.22 MDL-Nummer: MFCD00011303 MFCD00148878 InChI-Schlüssel: QCWXUUIWCKQGHC-UHFFFAOYSA-N Synonym: zircat,powder,circonio,zirconium, elemental,zirconio,zirkonium,compounds,unii-c6v6s92n3c,and compounds,foil PubChem CID: 23995 ChEBI: CHEBI:33342 IUPAC-Name: Zirkonium SMILES: [Zr]
Zirkonium-Standardlösung, für AAS, 1 mg/ml Zr in 5 % HF, Thermo Scientific Chemicals
CAS: 7440-67-7 | Molecular Formula: Zr | Molecular Weight: 91.22 g/mol
Zirkoniumfolie, 0.025 mm (0.001 Zoll) Dicke, geglüht, 99.8 % (Metallbasis), Thermo Scientific Chemicals
CAS: 7440-67-7 Summenformel: Zr Molekulargewicht (g/mol): 91.22 MDL-Nummer: MFCD00011303 MFCD00148878 InChI-Schlüssel: QCWXUUIWCKQGHC-UHFFFAOYSA-N Synonym: zircat,powder,circonio,zirconium, elemental,zirconio,zirkonium,compounds,unii-c6v6s92n3c,and compounds,foil PubChem CID: 23995 ChEBI: CHEBI:33342 IUPAC-Name: Zirkonium SMILES: [Zr]
Zirkoniumpulver, APS 2–3 Micron, Thermo Scientific Chemicals
CAS: 7440-67-7 Summenformel: Zr Molekulargewicht (g/mol): 91.22 MDL-Nummer: MFCD00011303 MFCD00148878 InChI-Schlüssel: QCWXUUIWCKQGHC-UHFFFAOYSA-N Synonym: zircat,powder,circonio,zirconium, elemental,zirconio,zirkonium,compounds,unii-c6v6s92n3c,and compounds,foil PubChem CID: 23995 ChEBI: CHEBI:33342 IUPAC-Name: Zirkonium SMILES: [Zr]
Zirkoniumdraht, 0.5 mm (0.020 Zoll) Durchmesser, geglüht, 99.2 % (Metallbasis ohne Hf) max. Hf 4.5 %, Thermo Scientific Chemicals
CAS: 7440-67-7 Summenformel: Zr Molekulargewicht (g/mol): 91.22 MDL-Nummer: MFCD00011303 MFCD00148878 InChI-Schlüssel: QCWXUUIWCKQGHC-UHFFFAOYSA-N Synonym: zircat,powder,circonio,zirconium, elemental,zirconio,zirkonium,compounds,unii-c6v6s92n3c,and compounds,foil PubChem CID: 23995 ChEBI: CHEBI:33342 IUPAC-Name: Zirkonium SMILES: [Zr]
Zirkoniumdraht, 1.0 mm (0.04 Zoll) Durchmesser, 99.2 % (Metallbasis ohne Hf), max. Hf 4.5 %, Thermo Scientific Chemicals
CAS: 7440-67-7 Summenformel: Zr Molekulargewicht (g/mol): 91.22 MDL-Nummer: MFCD00011303 MFCD00148878 InChI-Schlüssel: QCWXUUIWCKQGHC-UHFFFAOYSA-N Synonym: zircat,powder,circonio,zirconium, elemental,zirconio,zirkonium,compounds,unii-c6v6s92n3c,and compounds,foil PubChem CID: 23995 ChEBI: CHEBI:33342 IUPAC-Name: Zirkonium SMILES: [Zr]
Zirkoniumfolie, 0.711 mm (0.028 in.) Dicke, gebunden, 99.5 % (Metallbasis ohne Hf), Thermo Scientific Chemicals
CAS: 7440-67-7 Summenformel: Zr Molekulargewicht (g/mol): 91.22 MDL-Nummer: MFCD00011303 MFCD00148878 InChI-Schlüssel: QCWXUUIWCKQGHC-UHFFFAOYSA-N Synonym: zircat,powder,circonio,zirconium, elemental,zirconio,zirkonium,compounds,unii-c6v6s92n3c,and compounds,foil PubChem CID: 23995 ChEBI: CHEBI:33342 IUPAC-Name: Zirkonium SMILES: [Zr]
Zirkoniumdraht, 0.127 mm (0.005 Zoll) im Durchmesser, 99.95 % (Metallbasis ohne Hf), Thermo Scientific Chemicals
CAS: 7440-67-7 Summenformel: Zr Molekulargewicht (g/mol): 91.22 MDL-Nummer: MFCD00011303 MFCD00148878 InChI-Schlüssel: QCWXUUIWCKQGHC-UHFFFAOYSA-N Synonym: zircat,powder,circonio,zirconium, elemental,zirconio,zirkonium,compounds,unii-c6v6s92n3c,and compounds,foil PubChem CID: 23995 ChEBI: CHEBI:33342 IUPAC-Name: Zirkonium SMILES: [Zr]
Zirkoniumkugel, 3.175 mm (0.125 Zoll) Durchmesser x 3.175 mm (0.125 Zoll) Länge, ≥99.9 % (Metallbasis ohne Hf), nominales Hf 3 %, Thermo Scientific Chemicals
CAS: 7440-67-7 Summenformel: Zr Molekulargewicht (g/mol): 91.22 MDL-Nummer: MFCD00011303 MFCD00148878 InChI-Schlüssel: QCWXUUIWCKQGHC-UHFFFAOYSA-N Synonym: zircat,powder,circonio,zirconium, elemental,zirconio,zirkonium,compounds,unii-c6v6s92n3c,and compounds,foil PubChem CID: 23995 ChEBI: CHEBI:33342 IUPAC-Name: Zirkonium SMILES: [Zr]
Zirkoniumfolie, 0.25 mm (0.01 in.) Dicke, 99.94 % (Metallbasis ohne Hf), Thermo Scientific Chemicals
CAS: 7440-67-7 Summenformel: Zr Molekulargewicht (g/mol): 91.22 MDL-Nummer: MFCD00011303 MFCD00148878 InChI-Schlüssel: QCWXUUIWCKQGHC-UHFFFAOYSA-N Synonym: zircat,powder,circonio,zirconium, elemental,zirconio,zirkonium,compounds,unii-c6v6s92n3c,and compounds,foil PubChem CID: 23995 ChEBI: CHEBI:33342 IUPAC-Name: Zirkonium SMILES: [Zr]
Zirkoniumkugel, 3.175 mm (0.125 Zoll) Durchmesser x 6.35 mm (0.25 Zoll) Länge, 99.5 % (Metallbasis ohne Hf), nominales Hf 3 %, Thermo Scientific Chemicals
CAS: 7440-67-7 Summenformel: Zr Molekulargewicht (g/mol): 91.22 MDL-Nummer: MFCD00011303 MFCD00148878 InChI-Schlüssel: QCWXUUIWCKQGHC-UHFFFAOYSA-N Synonym: zircat,powder,circonio,zirconium, elemental,zirconio,zirkonium,compounds,unii-c6v6s92n3c,and compounds,foil PubChem CID: 23995 ChEBI: CHEBI:33342 IUPAC-Name: Zirkonium SMILES: [Zr]
Zirkoniumfolie, 0.25 mm (0.01 Zoll) Dicke, geglüht, 99.8 % (Metallbasis ohne Hf), Thermo Scientific Chemicals
CAS: 7440-67-7 Summenformel: Zr Molekulargewicht (g/mol): 91.22 MDL-Nummer: MFCD00011303 MFCD00148878 InChI-Schlüssel: QCWXUUIWCKQGHC-UHFFFAOYSA-N Synonym: zircat,powder,circonio,zirconium, elemental,zirconio,zirkonium,compounds,unii-c6v6s92n3c,and compounds,foil PubChem CID: 23995 ChEBI: CHEBI:33342 IUPAC-Name: Zirkonium SMILES: [Zr]
Zirkoniumkugel, 6.35 mm (0.25 Zoll) Durchmesser x 12.7 mm (0.50 Zoll) Länge, 99.5 % (Metallbasis ohne Hf), nominales Hf 3 %, Thermo Scientific Chemicals
CAS: 7440-67-7 Summenformel: Zr Molekulargewicht (g/mol): 91.22 MDL-Nummer: MFCD00011303 MFCD00148878 InChI-Schlüssel: QCWXUUIWCKQGHC-UHFFFAOYSA-N Synonym: zircat,powder,circonio,zirconium, elemental,zirconio,zirkonium,compounds,unii-c6v6s92n3c,and compounds,foil PubChem CID: 23995 ChEBI: CHEBI:33342 IUPAC-Name: Zirkonium SMILES: [Zr]
Zirkonium für AAS 1 g/L HNO₃ /HF-Matrix, Fisher chemical™
CAS: 7440-67-7 Summenformel: Zr Molekulargewicht (g/mol): 91.22 MDL-Nummer: MFCD00011303 MFCD00148878 InChI-Schlüssel: QCWXUUIWCKQGHC-UHFFFAOYSA-N PubChem CID: 23995 ChEBI: CHEBI:33342 IUPAC-Name: Zirkonium SMILES: [Zr]