CAS RN 75-52-5
Nitromethan, 99+ %, for analysis, Thermo Scientific Chemicals
CAS: 75-52-5 Summenformel: CH3NO2 Molekulargewicht (g/mol): 61.04 MDL-Nummer: MFCD00007400 InChI-Schlüssel: LYGJENNIWJXYER-UHFFFAOYSA-N Synonym: methane, nitro,nitrocarbol,nitrometan,nitrometan polish,nitro-methane,unii-ru5wg8c3f4,ccris 1205,hsdb 106,meno2,ru5wg8c3f4 PubChem CID: 6375 ChEBI: CHEBI:77701 IUPAC-Name: Nitromethan SMILES: C[N+]([O-])=O
Nitromethan, 96 %, for HPLC, Thermo Scientific Chemicals
CAS: 75-52-5 Summenformel: CH3NO2 Molekulargewicht (g/mol): 61.04 MDL-Nummer: MFCD00007400 InChI-Schlüssel: LYGJENNIWJXYER-UHFFFAOYSA-N Synonym: methane, nitro,nitrocarbol,nitrometan,nitrometan polish,nitro-methane,unii-ru5wg8c3f4,ccris 1205,hsdb 106,meno2,ru5wg8c3f4 PubChem CID: 6375 ChEBI: CHEBI:77701 IUPAC-Name: Nitromethan SMILES: C[N+]([O-])=O
Nitromethan, ≥98 %, Thermo Scientific Chemicals
CAS: 75-52-5 Summenformel: CH3NO2 Molekulargewicht (g/mol): 61.04 MDL-Nummer: MFCD00007400 InChI-Schlüssel: LYGJENNIWJXYER-UHFFFAOYSA-N Synonym: methane, nitro,nitrocarbol,nitrometan,nitrometan polish,nitro-methane,unii-ru5wg8c3f4,ccris 1205,hsdb 106,meno2,ru5wg8c3f4 PubChem CID: 6375 ChEBI: CHEBI:77701 IUPAC-Name: Nitromethan SMILES: C[N+]([O-])=O
Nitromethan, 99 %, Thermo Scientific Chemicals
CAS: 75-52-5 Summenformel: CH3NO2 Molekulargewicht (g/mol): 61.04 MDL-Nummer: MFCD00007400 InChI-Schlüssel: LYGJENNIWJXYER-UHFFFAOYSA-N Synonym: methane, nitro,nitrocarbol,nitrometan,nitrometan polish,nitro-methane,unii-ru5wg8c3f4,ccris 1205,hsdb 106,meno2,ru5wg8c3f4 PubChem CID: 6375 ChEBI: CHEBI:77701 IUPAC-Name: Nitromethan SMILES: C[N+]([O-])=O
-Nitromethan, 96 %, für Spektroskopie, Thermo Scientific Chemicals
CAS: 75-52-5 Summenformel: CH3NO2 Molekulargewicht (g/mol): 61.04 MDL-Nummer: MFCD00007400 InChI-Schlüssel: LYGJENNIWJXYER-UHFFFAOYSA-N Synonym: methane, nitro,nitrocarbol,nitrometan,nitrometan polish,nitro-methane,unii-ru5wg8c3f4,ccris 1205,hsdb 106,meno2,ru5wg8c3f4 PubChem CID: 6375 ChEBI: CHEBI:77701 IUPAC-Name: Nitromethan SMILES: C[N+]([O-])=O
Nitromethan, Reagenz ACS 95 %, Thermo Scientific Chemicals
CAS: 75-52-5 Summenformel: CH3NO2 Molekulargewicht (g/mol): 61.04 MDL-Nummer: MFCD00007400 InChI-Schlüssel: LYGJENNIWJXYER-UHFFFAOYSA-N Synonym: methane, nitro,nitrocarbol,nitrometan,nitrometan polish,nitro-methane,unii-ru5wg8c3f4,ccris 1205,hsdb 106,meno2,ru5wg8c3f4 PubChem CID: 6375 ChEBI: CHEBI:77701 IUPAC-Name: Nitromethan SMILES: C[N+]([O-])=O
Nitromethan, technisch, SLR, Fisher Chemical
CAS: 75-52-5 Summenformel: CH3NO2 Molekulargewicht (g/mol): 61.04 MDL-Nummer: MFCD00007400 InChI-Schlüssel: LYGJENNIWJXYER-UHFFFAOYSA-N Synonym: methane, nitro,nitrocarbol,nitrometan,nitrometan polish,nitro-methane,unii-ru5wg8c3f4,ccris 1205,hsdb 106,meno2,ru5wg8c3f4 PubChem CID: 6375 ChEBI: CHEBI:77701 IUPAC-Name: Nitromethan SMILES: C[N+]([O-])=O
Nitromethane, ≥99.0%, ReagentPlus™, Honeywell™
CAS: 75-52-5 Summenformel: CH3NO2 Molekulargewicht (g/mol): 61.04 MDL-Nummer: MFCD00007400 InChI-Schlüssel: LYGJENNIWJXYER-UHFFFAOYSA-N Synonym: methane, nitro,nitrocarbol,nitrometan,nitrometan polish,nitro-methane,unii-ru5wg8c3f4,ccris 1205,hsdb 106,meno2,ru5wg8c3f4 PubChem CID: 6375 ChEBI: CHEBI:77701 IUPAC-Name: Nitromethan SMILES: C[N+]([O-])=O