CAS RN 80451-05-4
CAS RN 80451-05-4
IUPAC Name:
—
Synonyme:
—
Molekulargewicht (g/mol):
—
Summenformel:
—
InChi Key:
—
SMILES:
—
1
–
3
von
3
Ergebnisse
1
MedChemExpress Cecropin B
MedChemExpress Cecropin B has high level of antimicrobial activity and is considered as a valuable peptide antibiotic.
2
MedChemExpress Cecropin B
MedChemExpress Cecropin B has high level of antimicrobial activity and is considered as a valuable peptide antibiotic.
3
MedChemExpress Cecropin B
MedChemExpress Cecropin B has high level of antimicrobial activity and is considered as a valuable peptide antibiotic.