CAS RN 81872-10-8
CAS RN 81872-10-8
IUPAC Name:
—
Synonyme:
—
Molekulargewicht (g/mol):
—
Summenformel:
—
InChi Key:
—
SMILES:
—
1
–
1
von
1
Ergebnisse
1
Zofenopril, MedChemExpress
MedChemExpress Zofenopril is an angiotensin-converting enzyme (ACE) inhibitor with an IC50 of 81 μM.