CAS RN 83373-60-8
CAS RN 83373-60-8
IUPAC Name:
tricyclo[5.2.1.0²,⁶]decan-8-yl sulfanylmethanethioate potassium
Synonyme:
tricyclo[5.2.1.0²,⁶]decan-8-yl sulfanylmethanethioate potassium
Molekulargewicht (g/mol):
267.47
Summenformel:
C11H16KOS2
InChi Key:
YFUFHKMDCZBHMY-UHFFFAOYNA-N
SMILES:
[K].SC(=S)OC1CC2CC1C1CCCC21
1
–
2
von
2
Ergebnisse
1
Tocris Bioscience™ D609
CAS: 83373-60-8 Summenformel: C11H18KOS2
2
D609, MedChemExpress
MedChemExpress D609, an antitumoural xanthate, is a specific and competitive phosphatidyl choline-specific phospholipase C (PC-PLC) inhibitor with a Ki of 6.4 μM. D609 is an antioxidative protector and has antiviral and anti-inflammatory activity.