CAS RN 890-98-2
IUPAC Name:
benzyl 2-hydroxy-2-phenylacetate
Synonyme:
benzyl 2-hydroxy-2-phenylacetate
Molekulargewicht (g/mol):
242.27
Summenformel:
C15H14O3
InChi Key:
JFKWZVQEMSKSBU-UHFFFAOYNA-N
SMILES:
OC(C(=O)OCC1=CC=CC=C1)C1=CC=CC=C1