CAS RN 90-89-1
CAS RN 90-89-1
IUPAC Name:
N,N-diethyl-4-methylpiperazine-1-carboxamide
Synonyme:
diethylcarbamazine camin carbamazine diethylcarbamazine citrate diethyl carbamazine hetrazan banocide notezine caracide caricide
Molekulargewicht (g/mol):
199.3
Summenformel:
C10H21N3O
InChi Key:
RCKMWOKWVGPNJF-UHFFFAOYSA-N
SMILES:
CCN(CC)C(=O)N1CCN(C)CC1
1
–
1
von
1
Ergebnisse
1
Thermo Scientific Chemicals Diethylcarbamazin
CAS: 90-89-1 Summenformel: C10H21N3O Molekulargewicht (g/mol): 199.30 InChI-Schlüssel: RCKMWOKWVGPNJF-UHFFFAOYSA-N IUPAC-Name: N,N-Diethyl-4-Methylpiperazin-1-Carboxamid SMILES: CCN(CC)C(=O)N1CCN(C)CC1