missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-[(tert-butoxycarbonyl)amino]cyclohexancarboxylsäure, 97 %, Thermo Scientific™
Click to view available options
Menge:
1 g
5 g
Spezifikation
Chemischer Name oder Material | 1-[(tert-Butoxycarbonyl)amino]cyclohexanecarboxylic acid |
CAS Min. % | 97.0 |
Prozentgehaltsbereich | 97% |
Formelmasse | 243.3 |
Reinheit (%) | 97% |
Menge | 1 g |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'