Learn More
ACROBiosystems Human B7-H7 / HHLA2 Protein, His Tag
Marke: ACROBiosystems B77-H52H5-100ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
B7-H7 (HHLA2) is a newly identified B7 family member that regulates human T-cell functions. B7-H7 was previously known as human endogenous retrovirus-H long terminal repeat associating 2 (HHLA2) with unidentified function. Recently, B7-H7 has been identified as a specific ligand for human CD28H. The B7-H7-CD28H pathway strongly promoted CD4+ T-cell proliferation and cytokine production via an AKT-dependent signaling cascade in the presence of TCR signaling, suggesting B7-H7 comprises a new co-stimulatory pathway. The first IgV domain of B7-H7, which presumably binds to a putative receptor, shows the highest homology to other B7 family members.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 38.9 kDa. The protein migrates as 48-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_009003.1 | |
100 μg | |
Ile 23 - Asn 344 | |
B7-H7,HHLA2,B7 Homolog 7 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |
38.9 kDa | |
Human B7-H7, His Tag (B77-H52H5) is expressed from human 293 cells (HEK293). It contains AA Ile 23 - Asn 344 (Accession # Q9UM44-1). | |
-20°C | |
B7-H7 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Immune Checkpoint Proteins |