Learn More
ACROBiosystems FITC-Labeled Human BCMA / TNFRSF17 Protein, His Tag DMF Filed
Marke: ACROBiosystems BCA-HF2H1-200ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Tumor necrosis factor receptor superfamily member 17 (TNFRSF17) is also known as B-cell maturation protein (BCMA), CD antigen CD269, which is a member of the TNF-receptor superfamily. TNFRSF17 contains one TNFR-Cys repeat. TNFRSF17 is expressed in mature B-cells, but not in T-cells or monocytes. TNFRSF17 is receptor for TNFSF13B/BLyS/BAFF and TNFSF13/APRIL. TNFRSF17 promotes B-cell survival and plays a role in the regulation of humoral immunity. TNFRSF17 can activate NF-kappa-B and JNK.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 7.8 kDa. The protein migrates as 10 kDa and 14-16 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_001183.2 | |
200 μg | |
Met 1 - Ala 54 | |
TNFRSF17,CD269,BCM,BCMA | |
FITC | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |
7.8 kDa | |
FITC-Labeled Human BCMA, His Tag (Cat. No. BCA-HF2H1) is expressed from human HEK293 cells. It contains AA Met 1 - Ala 54 (Accession # Q02223-1). It is the FITC labeled form of Human BCMA, His Tag (Cat. No. BCA-H522y). | |
-20°C, avoid light | |
BCMA | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Fluorescent labeled protein |