Learn More
Bevacizumab Mouse, Biotin, UltraLys, Monoclonal Antibody, AcroBiosystems
Mouse Monoclonal Antibody
Marke: ACROBiosystems BEB-BY13-200ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Bevacizumab Monoclonal antibody specifically detects Bevacizumab, recommended for PK/PD.Spezifikation
Bevacizumab | |
Biotin | |
200 μg | |
Primary |
Monoclonal | |
Hybridoma | |
RUO | |
-20°C |