missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spike RBD Neutralizing Mouse anti-SARS-CoV-2, Unconjugated, Clone: AM122, Chimeric mAb, AcroBiosystems
Mouse Monoclonal Antibody
Marke: ACROBiosystems SPD-M162-1mg
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Spike RBD Neutralizing Monoclonal antibody specifically detects Spike RBD Neutralizing in SARS-CoV-2 samples.Spezifikation
Spike RBD Neutralizing | |
AM122 | |
Human (HEK293) | |
RUO | |
SARS-CoV-2 | |
IgM |
Monoclonal | |
Unconjugated | |
1 mg | |
Primary | |
-20°C |