Learn More
DiscoverX InCELL Detection Kit
Click to view available options
Menge:
80 Plates, 8000/32000 dp (96-Well/384-Well)
Packungsgröße:
1 Set
Beschreibung
The InCELL Detection Kit is a detection kit intended for use with InCELL Hunter or Pulse cell lines, allowing measurement of target-engagement based changes in protein stability. This kit contains a lysis buffer and exogenous EA as well as EFC substrate to allow for detection of PL-tagged protein levels in the assayed cells. This kit contains all components required to assay 100 plates (96- or 384-well).
Spezifikation
Spezifikation
| Zur Verwendung mit (Anwendung) | Detection Kits |
| Menge | 80 Plates, 8000/32000 dp (96-Well/384-Well) |