missing translation for 'onlineSavingsMsg'
Learn More

2-Brom-5-Methylbenzoesäure, 98 %, Thermo Scientific Chemicals

Catalog Number p-7031747
Click to view available options
Menge:
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
6967-82-4
C8H7BrO2
215.046
MFCD00079722
ZXMISUUIYPFORW-UHFFFAOYSA-N
2-bromo-5-methyl-benzoic acid, 6-bromo-m-toluic acid, 2-bromo-5-methyl benzoic acid, benzoic acid, 2-bromo-5-methyl, 4-bromo-3-carboxytoluene, pubchem2166, acmc-1b7tl, ksc357q9r, 2-bromanyl-5-methyl-benzoic acid, 2-bromo-5-methylbenzoic acid
228080
2-Brom-5-Methylbenzoesäure
CC1=CC(=C(C=C1)Br)C(=O)O
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
6967-82-4
C8H7BrO2
215.046
MFCD00079722
ZXMISUUIYPFORW-UHFFFAOYSA-N
2-bromo-5-methyl-benzoic acid, 6-bromo-m-toluic acid, 2-bromo-5-methyl benzoic acid, benzoic acid, 2-bromo-5-methyl, 4-bromo-3-carboxytoluene, pubchem2166, acmc-1b7tl, ksc357q9r, 2-bromanyl-5-methyl-benzoic acid, 2-bromo-5-methylbenzoic acid
228080
2-Brom-5-Methylbenzoesäure
CC1=CC(=C(C=C1)Br)C(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 2-Bromo-5-methylbenzoic acid
Menge 1 g
Formelmasse 215.05
Reinheit (%) 98%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'