missing translation for 'onlineSavingsMsg'
Learn More

2-Chlor-5-methylbenzoesäure, 98 %, Thermo Scientific™

Catalog Number p-7028182
Click to view available options
Menge:
1 g
5 g
25 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
6342-60-5
C8H7ClO2
170.592
MFCD00045798
LEBWXJZAWTVKFL-UHFFFAOYSA-N
2-chloro-5-methylbenzole acid, 6-chloro-m-toluic acid, benzoic acid, 2-chloro-5-methyl, pubchem12501, 3-carboxy-4-chlorotoluene, acmc-1b72o, ksc497q1d, 2-chloro-5-methylbenozic acid, 2-chloro-5-methyl-benzoic acid, lebwxjzawtvkfl-uhfffaoysa
240430
2-Chlor-5-methylbenzoesäure
CC1=CC(=C(C=C1)Cl)C(=O)O
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
6342-60-5
C8H7ClO2
170.592
MFCD00045798
LEBWXJZAWTVKFL-UHFFFAOYSA-N
2-chloro-5-methylbenzole acid, 6-chloro-m-toluic acid, benzoic acid, 2-chloro-5-methyl, pubchem12501, 3-carboxy-4-chlorotoluene, acmc-1b72o, ksc497q1d, 2-chloro-5-methylbenozic acid, 2-chloro-5-methyl-benzoic acid, lebwxjzawtvkfl-uhfffaoysa
240430
2-Chlor-5-methylbenzoesäure
CC1=CC(=C(C=C1)Cl)C(=O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 2-Chloro-5-methylbenzoic acid
Menge 1 g
Formelmasse 170.6
Reinheit (%) 98%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'