missing translation for 'onlineSavingsMsg'
Learn More

2-Chlor-N-(2-Methylphenyl)-4-Nitrobenzamid, 97 %, Thermo Scientific™

Catalog Number 15466038
Click to view available options
Menge:
250 mg
1 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
329940-78-5
C14H11ClN2O3
290.703
MFCD01188644
XQBCDIDTLUWUMG-UHFFFAOYSA-N
2-chloro-n-2-methylphenyl-4-nitrobenzamide, 2-chloro-4-nitro-n-o-tolyl-benzamide, 2-chloro-4-nitrophenyl-n-2-methylphenyl carboxamide
710907
2-Chlor-N-(2-methylphenyl)-4-nitrobenzamid
CC1=CC=CC=C1NC(=O)C2=C(C=C(C=C2)[N+](=O)[O-])Cl
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
329940-78-5
C14H11ClN2O3
290.703
MFCD01188644
XQBCDIDTLUWUMG-UHFFFAOYSA-N
2-chloro-n-2-methylphenyl-4-nitrobenzamide, 2-chloro-4-nitro-n-o-tolyl-benzamide, 2-chloro-4-nitrophenyl-n-2-methylphenyl carboxamide
710907
2-Chlor-N-(2-methylphenyl)-4-nitrobenzamid
CC1=CC=CC=C1NC(=O)C2=C(C=C(C=C2)[N+](=O)[O-])Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 2-Chloro-N-(2-methylphenyl)-4-nitrobenzamide
Menge 1 g
Formelmasse 290.7
Reinheit (%) 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'