missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-5-(Pentafluorothio)Benzoesäure, 97 %

Catalog Number p-7031847
Click to view available options
Menge:
250 mg
1 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
623943-54-4
C8H7F5O2S
262.194
MFCD16652477
HSIPIUUYFOAFKK-UHFFFAOYSA-N
2-methyl-5-pentafluorosulfur benzoic acid, 2-methyl-5-pentafluoro-??-sulfanyl benzoic acid, 2-methyl-5-pentafluorothio benzoic acid
66523588
2-methyl-5-(pentafluor-$l^{6}-sulfanyl)benzoesäure
CC1=C(C=C(C=C1)S(F)(F)(F)(F)F)C(=O)O
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
623943-54-4
C8H7F5O2S
262.194
MFCD16652477
HSIPIUUYFOAFKK-UHFFFAOYSA-N
2-methyl-5-pentafluorosulfur benzoic acid, 2-methyl-5-pentafluoro-??-sulfanyl benzoic acid, 2-methyl-5-pentafluorothio benzoic acid
66523588
2-methyl-5-(pentafluor-$l^{6}-sulfanyl)benzoesäure
CC1=C(C=C(C=C1)S(F)(F)(F)(F)F)C(=O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 2-Methyl-5-(pentafluorothio)benzoic acid
Menge 250 mg
Formelmasse 262.19
Reinheit (%) 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'