Learn More
Hamilton™ 250 μL 700 Series Microliter Syringe
The 700 Series Microliter Syringe fulfills the general liquid handling requirements of manufacturing and research laboratories.
Marke: Hamilton™ CAL80765
Weitere Details : Netto-Gewicht : 0.03300kg
Beschreibung
- The 700 series is the original Hamilton syringe.
- This series of Microliter syringes is designed to solve the general liquid handling requirements of manufacturing and research laboratories.
- The 700 series remains the industry standard.
- The plungers and barrels are not interchangeable or replaceable.
- Dispenses volumes up to 250 μL.
Spezifikation
Autoclavable | |
26s G | |
0.005 in. | |
0.019 in. | |
1.24 mm | |
0.474 mm | |
239°F | |
50°F | |
Cone | |
1 | |
43.4 mm |
Stainless Steel | |
0.049 in. | |
0.13 mm | |
0.260 in. | |
6.60 mm | |
1.71 in. | |
115°C | |
7000.5 KH | |
10°C | |
0.5 μL | |
Knurled Hub |