missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,3',5,5'-Tetramethylbenzidinlösung, gebrauchsfertig, fällt aus, hohe Empfindlichkeit, Thermo Scientific Chemicals
Click to view available options
Menge:
125 mL
500 mL
1000 mL
Beschreibung
Excellent colorimetric substrate for detection of horseradish peroxidase labelled probes and is used with peroxidase and peroxidase coupled systems, particularly in ELISA techniques, blue result
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Ausgezeichnetes kolorimetrisches Substrat für den Nachweis von mit Meerrettich-Peroxidase markierten Sonden, zudem Verwendung mit Peroxidase- und Peroxidase-gekoppelten Systemen, insbesondere in ELISA-Verfahren, blaues Resultat
Spezifikation
Chemischer Name oder Material | 3,3',5,5'-Tetramethylbenzidine Solution |
Namenshinweis | Ready-to-Use, precipitating, high sensitivity |
Farbe | Farblos bis hellgelb |
Geruch | Alcohol-like |
Menge | 125 mL |
UN-Nummer | UN2810 |
Beilstein | 2808541 |
Formelmasse | 240.35 |
Physikalische Form | Flüssig |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'