missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-[4-(Tert-butoxycarbonyl)-1,4-diazepan-1-yl]propansäure, Maybridge
Click to view available options
Menge:
1 g
Verpackung:
Bernsteinfarbene Glasflasche
Spezifikation
Chemischer Name oder Material | 3-[4-(tert-butoxycarbonyl)-1, 4-diazepan-1-yl]propanoic acid |
CAS | 874801-64-6 |
Summenformel | C13H24N2O4 |
Verpackung | Bernsteinfarbene Glasflasche |
Synonym | 3-4-tert-butoxycarbonyl-1,4-diazepan-1-yl propanoic acid, 3-4-tert-butoxy carbonyl-1,4-diazepan-1-yl propanoic acid, 4-2-carboxy-ethyl-1,4 diazepane-1-carboxylic acid tert-butyl ester |
InChI-Schlüssel | QXUVNKBILZAHRW-UHFFFAOYSA-N |
SMILES | CC(C)(C)OC(=O)N1CCCN(CC1)CCC(=O)O |
IUPAC-Name | 3-[4-[(2-Methylpropan-2-yl)oxycarbonyl]-1,4-diazepan-1-yl]propansäure |
Molekulargewicht (g/mol) | 272.345 |
PubChem CID | 18526454 |
Mehr anzeigen |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'