missing translation for 'onlineSavingsMsg'
Learn More

3-Fluor-4-Methoxybenzoesäure, 98 %, Thermo Scientific™

Catalog Number p-7026109
Click to view available options
Menge:
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
403-20-3
C8H6FO3
169.13
MFCD00060675
HYNNNQDQEORWEU-UHFFFAOYSA-M
3-fluoro-4-methoxy-benzoic acid, 3-fluoro-p-anisic acid, 3-fluoro-4-methoxy benzoic acid, 3-fluoro-4-methoxybenzoicacid, pubchem1393, opera_id_903, acmc-209jd1, ksc494s2f, 3-fluoro-4-methoxy benzoicacid, rarechem al bo 0870
2733401
3-fluoro-4-methoxybenzoate
COC1=CC=C(C=C1F)C([O-])=O
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
403-20-3
C8H6FO3
169.13
MFCD00060675
HYNNNQDQEORWEU-UHFFFAOYSA-M
3-fluoro-4-methoxy-benzoic acid, 3-fluoro-p-anisic acid, 3-fluoro-4-methoxy benzoic acid, 3-fluoro-4-methoxybenzoicacid, pubchem1393, opera_id_903, acmc-209jd1, ksc494s2f, 3-fluoro-4-methoxy benzoicacid, rarechem al bo 0870
2733401
3-fluoro-4-methoxybenzoate
COC1=CC=C(C=C1F)C([O-])=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 3-Fluoro-4-methoxybenzoic acid
Schmelzpunkt 209°C to 213°C
Menge 1 g
Beilstein 2087583
Formelmasse 170.14
Reinheit (%) 98%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'