missing translation for 'onlineSavingsMsg'
Learn More

3'-Methoxybiphenyl-3-Carbonsäure, 95 %, Thermo Scientific™

Catalog Number p-7034113
Click to view available options
Menge:
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
168618-45-9
C14H11O3
227.24
MFCD03424598
JFPVSZXZHHOWMI-UHFFFAOYSA-M
3'-methoxybiphenyl-3-carboxylic acid, 3-3-methoxyphenyl benzoic acid, 3'-methoxy-1,1'-biphenyl-3-carboxylic acid, 3'-methoxy-biphenyl-3-carboxylic acid, 3-biphenyl-3'-methoxy carboxylic acid, 3'-methoxy 1,1'-biphenyl-3-carboxylic acid, acmc-1c1el, amtda021, 3'-methoxy-3-biphenylcarboxylic acid, 3'-methoxy-1,1'-biphenyl-3-carboxylicacid
2759550
3-(3-methoxyphenyl)benzoesäure
COC1=CC=CC(=C1)C1=CC=CC(=C1)C([O-])=O
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
168618-45-9
C14H11O3
227.24
MFCD03424598
JFPVSZXZHHOWMI-UHFFFAOYSA-M
3'-methoxybiphenyl-3-carboxylic acid, 3-3-methoxyphenyl benzoic acid, 3'-methoxy-1,1'-biphenyl-3-carboxylic acid, 3'-methoxy-biphenyl-3-carboxylic acid, 3-biphenyl-3'-methoxy carboxylic acid, 3'-methoxy 1,1'-biphenyl-3-carboxylic acid, acmc-1c1el, amtda021, 3'-methoxy-3-biphenylcarboxylic acid, 3'-methoxy-1,1'-biphenyl-3-carboxylicacid
2759550
3-(3-methoxyphenyl)benzoesäure
COC1=CC=CC(=C1)C1=CC=CC(=C1)C([O-])=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 3'-Methoxybiphenyl-3-carboxylic acid
Menge 1 g
Formelmasse 228.25
Reinheit (%) 95%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'