missing translation for 'onlineSavingsMsg'
Learn More

3-Nitrobenzylaminhydrochlorid, 97+ %, Thermo Scientific Chemicals

Catalog Number p-7026965
Click to view available options
Menge:
1 g
5 g
25 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
26177-43-5
C7H9ClN2O2
188.611
MFCD00012858
DLZXLCHQWOZGSE-UHFFFAOYSA-N
3-nitrobenzylamine hydrochloride, 3-nitrophenyl methanamine hydrochloride, 3-nitrobenzylamine hcl, 3-nitrobenzylammonium hydrochloride, benzenemethanamine, 3-nitro-, monohydrochloride, 1-3-nitrophenyl methanamine hydrochloride, pubchem7485, chembl13978, m-nitrobenzylamine hydrochloride, 3-nitrobenzyl amine hydrochloride
2724030
(3-nitrophenyl)methanamin;hydrochlorid
C1=CC(=CC(=C1)[N+](=O)[O-])CN.Cl
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
26177-43-5
C7H9ClN2O2
188.611
MFCD00012858
DLZXLCHQWOZGSE-UHFFFAOYSA-N
3-nitrobenzylamine hydrochloride, 3-nitrophenyl methanamine hydrochloride, 3-nitrobenzylamine hcl, 3-nitrobenzylammonium hydrochloride, benzenemethanamine, 3-nitro-, monohydrochloride, 1-3-nitrophenyl methanamine hydrochloride, pubchem7485, chembl13978, m-nitrobenzylamine hydrochloride, 3-nitrobenzyl amine hydrochloride
2724030
(3-nitrophenyl)methanamin;hydrochlorid
C1=CC(=CC(=C1)[N+](=O)[O-])CN.Cl

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 3-Nitrobenzylamine hydrochloride
Schmelzpunkt ∼225°C (decomposition)
Menge 1 g
Formelmasse 188.62
Reinheit (%) 97%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'