missing translation for 'onlineSavingsMsg'
Learn More

4-[3-(Trifluoromethyl)phenylsulfonylaminomethyl]Benzoesäure, 96 %, Thermo Scientific™

Catalog Number p-7031918
Click to view available options
Menge:
250 mg
1 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
690645-93-3
C15H12F3NO4S
359.319
MFCD04154032
YULRKVYOEFOBSH-UHFFFAOYSA-N
4-3-trifluoromethyl phenyl sulfonamido methyl benzoic acid, 4-3-trifluoromethyl phenylsulfonamido methyl benzoic acid, 4-3-trifluoromethyl phenylsulfonylaminomethyl benzoic acid, 4-3-trifluoromethyl benzenesulfonamido methyl benzoic acid, 4-3-trifluoromethyl phenyl sulfonylamino methyl benzoic acid
984839
4-[[[3-(trifluormethyl)phenyl]sulfonylamino]methyl]benzoesäure
C1=CC(=CC(=C1)S(=O)(=O)NCC2=CC=C(C=C2)C(=O)O)C(F)(F)F
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
690645-93-3
C15H12F3NO4S
359.319
MFCD04154032
YULRKVYOEFOBSH-UHFFFAOYSA-N
4-3-trifluoromethyl phenyl sulfonamido methyl benzoic acid, 4-3-trifluoromethyl phenylsulfonamido methyl benzoic acid, 4-3-trifluoromethyl phenylsulfonylaminomethyl benzoic acid, 4-3-trifluoromethyl benzenesulfonamido methyl benzoic acid, 4-3-trifluoromethyl phenyl sulfonylamino methyl benzoic acid
984839
4-[[[3-(trifluormethyl)phenyl]sulfonylamino]methyl]benzoesäure
C1=CC(=CC(=C1)S(=O)(=O)NCC2=CC=C(C=C2)C(=O)O)C(F)(F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 4-[3-(Trifluoromethyl)phenylsulfonylaminomethyl]benzoic acid
Schmelzpunkt 219°C to 220°C
Menge 250 mg
Formelmasse 359.33
Reinheit (%) 96%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'