missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(4-Cyanophenoxy)benzoesäure, 97 %, Thermo Scientific™
Click to view available options
Menge:
1 g
Spezifikation
Chemischer Name oder Material | 4-(4-Cyanophenoxy)benzoic acid |
CAS | 50793-29-8 |
Prozentgehaltsbereich | 97% |
Summenformel | C14H9NO3 |
Menge | 1 g |
Synonym | 4-4-cyanophenoxy benzoic acid, 4-4-cyano-phenoxy-benzoic acid, cambridge id 5128427, 4-4'-cyanophenoxy benzoic acid |
InChI-Schlüssel | JBBCZYHRHADVJB-UHFFFAOYSA-N |
SMILES | C1=CC(=CC=C1C#N)OC2=CC=C(C=C2)C(=O)O |
IUPAC-Name | 4-(4-Dimethylaminophenoxy)Benzoesäure |
Molekulargewicht (g/mol) | 239.23 |
Mehr anzeigen |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'