missing translation for 'onlineSavingsMsg'
Learn More

4-Chlor-2-Fluor-3-Methoxyphenylessigsäure, 97 %

Catalog Number p-7032084
Click to view available options
Menge:
250 mg
1 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1323955-64-1
C9H8ClFO3
218.608
MFCD19687184
ISTHAWLIXVWDQR-UHFFFAOYSA-N
4-chloro-2-fluoro-3-methoxyphenylacetic acid, 4-chloro-2-fluoro-3-methoxyphenyl acetic acid, 2-4-chloro-2-fluoro-3-methoxyphenyl acetic acid
66523613
2-(4-Chlor-2-Fluor-3-methoxyphenyl)essigsäure
COC1=C(C=CC(=C1F)CC(=O)O)Cl
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1323955-64-1
C9H8ClFO3
218.608
MFCD19687184
ISTHAWLIXVWDQR-UHFFFAOYSA-N
4-chloro-2-fluoro-3-methoxyphenylacetic acid, 4-chloro-2-fluoro-3-methoxyphenyl acetic acid, 2-4-chloro-2-fluoro-3-methoxyphenyl acetic acid
66523613
2-(4-Chlor-2-Fluor-3-methoxyphenyl)essigsäure
COC1=C(C=CC(=C1F)CC(=O)O)Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 4-Chloro-2-fluoro-3-methoxyphenylacetic acid
Menge 250 mg
Formelmasse 218.61
Reinheit (%) 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'