missing translation for 'onlineSavingsMsg'
Learn More

4-Chlor-3-(Methylcarbamoyl)benzenboronsäure, 98 %, Thermo Scientific™

Catalog Number p-7034957
Click to view available options
Menge:
250 mg
1 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
871332-65-9
C8H9BClNO3
213.424
MFCD07363795
SYZZWFMATUWETK-UHFFFAOYSA-N
4-chloro-3-n-methylcarbamoyl phenylboronic acid, 4-chloro-3-methylcarbamoyl phenyl boronic acid, 4-chloro-3-methylcarbamoyl phenylboronic acid, n-methyl 2-chloro-5-boronobenzamide, n-methyl 5-borono-2-chlorobenzamide, boronic acid,b-4-chloro-3-methylamino carbonyl phenyl, b-4-chloro-3-methylamino carbonyl phenyl-boronic acid, boronic acid, b-4-chloro-3-methylamino carbonyl phenyl, acmc-209qgv
44886929
[4-chlor-3-(methylcarbamoyl)phenyl]boronsäure
B(C1=CC(=C(C=C1)Cl)C(=O)NC)(O)O
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
871332-65-9
C8H9BClNO3
213.424
MFCD07363795
SYZZWFMATUWETK-UHFFFAOYSA-N
4-chloro-3-n-methylcarbamoyl phenylboronic acid, 4-chloro-3-methylcarbamoyl phenyl boronic acid, 4-chloro-3-methylcarbamoyl phenylboronic acid, n-methyl 2-chloro-5-boronobenzamide, n-methyl 5-borono-2-chlorobenzamide, boronic acid,b-4-chloro-3-methylamino carbonyl phenyl, b-4-chloro-3-methylamino carbonyl phenyl-boronic acid, boronic acid, b-4-chloro-3-methylamino carbonyl phenyl, acmc-209qgv
44886929
[4-chlor-3-(methylcarbamoyl)phenyl]boronsäure
B(C1=CC(=C(C=C1)Cl)C(=O)NC)(O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 4-Chloro-3-(methylcarbamoyl)benzeneboronic acid
Schmelzpunkt 294°C to 298°C
Menge 250 mg
Formelmasse 213.43
Reinheit (%) 98%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'