missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxy-2-(pentafluorthio)-Benzoesäure, 98 %, Thermo Scientific™

Click to view available options
Menge:
250 mg
1 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
127817-85-0
C9H7F3O3
220.15
MFCD01091013
MLCRFQSWSHKLDP-UHFFFAOYSA-N
4-methoxy-2-trifluoromethyl benzoic acid, benzoic acid, 4-methoxy-2-trifluoromethyl, pubchem4515, 4-methoxy-2-trifluoromethyl benzoicacid, acmc-20a4b9, ksc515o4d, rarechem al be 1192, 2-trifluoromethyl-p-anisic acid, 2-carboxy-5-methoxybenzotrifluoride, 4-carboxy-3-trifluoromethyl anisole
2775294
4-methoxy-2-(trifluormethyl)benzoesäure
COC1=CC(=C(C=C1)C(O)=O)C(F)(F)F
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
127817-85-0
C9H7F3O3
220.15
MFCD01091013
MLCRFQSWSHKLDP-UHFFFAOYSA-N
4-methoxy-2-trifluoromethyl benzoic acid, benzoic acid, 4-methoxy-2-trifluoromethyl, pubchem4515, 4-methoxy-2-trifluoromethyl benzoicacid, acmc-20a4b9, ksc515o4d, rarechem al be 1192, 2-trifluoromethyl-p-anisic acid, 2-carboxy-5-methoxybenzotrifluoride, 4-carboxy-3-trifluoromethyl anisole
2775294
4-methoxy-2-(trifluormethyl)benzoesäure
COC1=CC(=C(C=C1)C(O)=O)C(F)(F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemischer Name oder Material 4-Methoxy-2-(trifluoromethyl)benzoic acid
Schmelzpunkt 147°C to 152°C
Menge 250 mg
Formelmasse 220.15
Reinheit (%) 98%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'