Learn More
4-Methoxycarbonyl-2-Nitrobenzenboronsäure, 97 %, Thermo Scientific Chemicals
Catalog Number p-7035149
Click to view available options
Menge:
1 g
5 g
85107-55-7
C8H8BNO6
224.963
MFCD01632202
SFEJGHJCGQNFQC-UHFFFAOYSA-N
4-methoxycarbonyl-2-nitrophenyl boronic acid, 4-methoxycarbonyl-2-nitrophenylboronic acid, 4-methoxycarbonyl-2-nitrobenzeneboronic acid, methyl 4-dihydroxyboranyl-3-nitrobenzoate, 2-nitro-4-methoxycarbonyl phenylboronic acid, 4-methoxycarbonyl-2-nitro-phenyl boronic acid, methyl 4-borono-3-nitrobenzoate, acmc-209q4d
2773493
(4-methoxycarbonyl-2-nitrophenyl)boronsäure
B(C1=C(C=C(C=C1)C(=O)OC)[N+](=O)[O-])(O)O
85107-55-7
C8H8BNO6
224.963
MFCD01632202
SFEJGHJCGQNFQC-UHFFFAOYSA-N
4-methoxycarbonyl-2-nitrophenyl boronic acid, 4-methoxycarbonyl-2-nitrophenylboronic acid, 4-methoxycarbonyl-2-nitrobenzeneboronic acid, methyl 4-dihydroxyboranyl-3-nitrobenzoate, 2-nitro-4-methoxycarbonyl phenylboronic acid, 4-methoxycarbonyl-2-nitro-phenyl boronic acid, methyl 4-borono-3-nitrobenzoate, acmc-209q4d
2773493
(4-methoxycarbonyl-2-nitrophenyl)boronsäure
B(C1=C(C=C(C=C1)C(=O)OC)[N+](=O)[O-])(O)O
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 4-Methoxycarbonyl-2-nitrobenzeneboronic acid
Schmelzpunkt 160°C to 171°C
Menge 1 g
Formelmasse 224.96
Reinheit (%) 97%