Learn More
ABL1 [T315I] Recombinant Human Protein, Invitrogen™
The T315I mutant is a Gleevec resistant form of ABL1.
Marke: Invitrogen PV4184
Weitere Details : Netto-Gewicht : 0.00900kg
Beschreibung
The ABL1 proto-oncogene encodes a cytoplasmic and nuclear protein tyrosine kinase that has been implicated in processes of cell differentiation, cell division, cell adhesion, and stress response.
Spezifikation
NP_005148.2, T315I | |
50mM Tris-HCl, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% NP-40, 2mM DTT and 50% glycerol | |
1mg | |
RUO - research use only | |
ABL1 T315I, ABL, c-ABL | |
Human | |
His Tag | |
Frozen |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
125.4kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Full Length | |
ABL Kinase Family | |
Recombinant | |
Baculovirus |