missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human ADRBK1 (GRK2), His Tag Recombinant Protein
Human ADRBK1 (GRK2), His Tag Recombinant Protein
Marke: Invitrogen™ PR4694A
Weitere Details : Netto-Gewicht : 0.00900kg
Beschreibung
ADRBK1 (GRK2) is a serine⁄threonine kinase. Many of the G protein-coupled receptors are known to be phosphorylated by G protein-dependent receptor kinases (GRKs)Spezifikation
NP_001610.2 | |
Protein Assays and Analysis, Kinase Assay | |
82.3kDa | |
1mg | |
For research use only. Not for use in diagnostic procedures | |
ADRBK1 (GRK2) | |
ADRBK1 (GRK2) was pre-diluted in enzyme dilution buffer and assayed in 25mM Tris (pH 7.5), 10mM MgCl2, 0.5mM EGTA, 0.5mM Na3VO4, 5mM β−glycerophosphate, 2.5mM DTT, 0.01% Triton™ X-100, 200μM ATP, 1000 μg/mL Tubulin and trace [32P]-γ-ATP for 10 minutes at 30°C | |
ADRBK1 (GRK2) Recombinant Human Protein | |
Human | |
Baculovirus | |
95% by SDS-PAGE |
0.49mg/mL | |
Frozen | |
ADRBK1 (GRK2) Recombinant Human Protein | |
–80°C | |
1 Each | |
ADRBK1 (GRK2) Recombinant Human Protein | |
GRK kinase family | |
58nmoles of phosphate transferred to 0000 Place Holder - per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 1.25μg/mL | |
His Tag | |
Frozen |
For Research Use Only. Not for use in diagnostic procedures.