missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AKT2 (PKB beta) Recombinant Human Protein
AKT2 is a phosphoinositide-dependent serine/threonine protein kinase that is enriched in insulin responsive tissues.
Marke: Invitrogen™ PV3975
Weitere Details : Netto-Gewicht : 0.00900kg
Beschreibung
AKT2 has been implicated in the metabolic actions of the hormone.
Spezifikation
NP_001617 | |
20mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.01% Triton X-100, 0.1mM Na3VO4, 2mM DTT, 20% glycerol | |
1mg | |
RUO - research use only | |
AKT2 (PKB beta) | |
Human | |
His Tag | |
Frozen |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
59.1kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Full Length | |
AKT Kinase Family | |
Recombinant | |
Baculovirus |